accessibility menu, dialog, popup

UserName

CH3CH3NONOO

CAS RN 37699-43-7

IUPAC Name: 2,3-dimethyl-4-nitropyridin-1-ium-1-olate
Synonyms: 2,3-dimethyl-4-nitropyridin-1-ium-1-olate
Molecular Weight (g/mol): 168.15
Molecular Formula: C7H8N2O3
InChi Key: CFMTVTYBZMKULI-UHFFFAOYSA-N
SMILES: CC1=C(C)[N+]([O-])=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)