missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-4-nitropyridine N-Oxide 98.0+%, TCI America™

Catalog No. D290525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D290525G 25 g
D29055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D290525G Supplier TCI America Supplier No. D290525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 37699-43-7
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.15
MDL Number MFCD00065172
InChI Key CFMTVTYBZMKULI-UHFFFAOYSA-N
Synonym 2,3-dimethyl-4-nitropyridine-n-oxide, 2,3-dimethyl-4-nitropyridine 1-oxide, 2,3-dimethyl-4-nitropyridine n-oxide, 4-nitro-2,3-lutidine-n-oxide, pyridine, 2,3-dimethyl-4-nitro-, 1-oxide, 2,3-dimethyl-4-nitropyridin-1-ium-1-olate, 4-nitro-2,3-lutidine n-oxide, 4-nitro-2,3-dimethylpyridine n-oxide, 2,3-dimethyl-4-nitro-pyridine 1-oxide, 2,3-dimethyl-4-nitro-1-oxido-pyridin-1-ium
PubChem CID 148223
IUPAC Name 2,3-dimethyl-4-nitropyridin-1-ium-1-olate
SMILES CC1=C(C)[N+]([O-])=CC=C1[N+]([O-])=O

Specifications

Melting Point 92°C
Quantity 25 g
Formula Weight 168.15
Percent Purity ≥98.0% (T)
Grade Reagent
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dimethyl-4-nitropyridine N-Oxide
RTECSNumber UT2807000
TSCA No