missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-4-nitrophenol 98.0+%, TCI America™

Catalog No. D26085G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26085G 5 g
D260825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26085G Supplier TCI America Supplier No. D26085G

Chemical Identifiers

CAS 19499-93-5
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
MDL Number MFCD01861284
InChI Key DAEZHJNJHQAEPE-UHFFFAOYSA-N
Synonym 4-Nitro-2,3-xylenol
PubChem CID 13331245
IUPAC Name 2,3-dimethyl-4-nitrophenol
SMILES CC1=C(O)C=CC(=C1C)[N+]([O-])=O

Specifications

Melting Point 125°C
Quantity 5 g
Formula Weight 167.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dimethyl-4-nitrophenol