missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dimethyl-1-propylimidazolium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
$331.81 - $995.41
Chemical Identifiers
| CAS | 169051-76-7 |
|---|---|
| Molecular Formula | C10H15F6N3O4S2 |
| Molecular Weight (g/mol) | 419.357 |
| MDL Number | MFCD03788922 |
| InChI Key | XOZHIVUWCICHSQ-UHFFFAOYSA-N |
| Synonym | 1,2-Dimethyl-3-propylimidazolium Bis(trifluoromethanesulfonyl)imide |
| PubChem CID | 11750474 |
| IUPAC Name | bis(trifluoromethylsulfonyl)azanide;1,2-dimethyl-3-propylimidazol-1-ium |
| SMILES | CCCN1C=C[N+](=C1C)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F |
Chemical Identifiers
| 169051-76-7 | |
| 419.357 | |
| XOZHIVUWCICHSQ-UHFFFAOYSA-N | |
| 11750474 | |
| CCCN1C=C[N+](=C1C)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F |
| C10H15F6N3O4S2 | |
| MFCD03788922 | |
| 1,2-Dimethyl-3-propylimidazolium Bis(trifluoromethanesulfonyl)imide | |
| bis(trifluoromethylsulfonyl)azanide;1,2-dimethyl-3-propylimidazol-1-ium |
Specifications
| 169051-76-7 | |
| Yellow | |
| MFCD03788922 | |
| 1,2-Dimethyl-3-propylimidazolium Bis(trifluoromethanesulfonyl)imide | |
| CCCN1C=C[N+](=C1C)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F | |
| 419.357 | |
| 419.36 | |
| Liquid |
| 8°C | |
| C10H15F6N3O4S2 | |
| 5 g | |
| XOZHIVUWCICHSQ-UHFFFAOYSA-N | |
| bis(trifluoromethylsulfonyl)azanide;1,2-dimethyl-3-propylimidazol-1-ium | |
| 11750474 | |
| ≥98.0% (T) | |
| 2,3-Dimethyl-1-propylimidazolium Bis(trifluoromethanesulfonyl)imide |
Safety and Handling
TSCA : No