CAS RN 169051-76-7
IUPAC Name:
2,3-dimethyl-1-propyl-1H-imidazol-3-ium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms:
1,2-dimethyl-3-propylimidazol-1-ium; bistriflylimide anion
Molecular Weight (g/mol):
419.36
Molecular Formula:
C10H15F6N3O4S2
InChi Key:
XOZHIVUWCICHSQ-UHFFFAOYSA-N
SMILES:
CCCN1C=C[N+](C)=C1C.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
1
–
1
of
1
results
1
2,3-Dimethyl-1-propylimidazolium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
CAS: 169051-76-7 Molecular Formula: C10H15F6N3O4S2 Molecular Weight (g/mol): 419.357 MDL Number: MFCD03788922 InChI Key: XOZHIVUWCICHSQ-UHFFFAOYSA-N Synonym: 1,2-Dimethyl-3-propylimidazolium Bis(trifluoromethanesulfonyl)imide PubChem CID: 11750474 IUPAC Name: bis(trifluoromethylsulfonyl)azanide;1,2-dimethyl-3-propylimidazol-1-ium SMILES: CCCN1C=C[N+](=C1C)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F