missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-1-propylimidazolium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. D42895G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42895G 5 g
D428925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42895G Supplier TCI America Supplier No. D42895G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 169051-76-7
Molecular Formula C10H15F6N3O4S2
Molecular Weight (g/mol) 419.357
MDL Number MFCD03788922
InChI Key XOZHIVUWCICHSQ-UHFFFAOYSA-N
Synonym 1,2-Dimethyl-3-propylimidazolium Bis(trifluoromethanesulfonyl)imide
PubChem CID 11750474
IUPAC Name bis(trifluoromethylsulfonyl)azanide;1,2-dimethyl-3-propylimidazol-1-ium
SMILES CCCN1C=C[N+](=C1C)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point 8°C
Color Yellow
Quantity 5 g
Formula Weight 419.36
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2,3-Dimethyl-1-propylimidazolium Bis(trifluoromethanesulfonyl)imide