missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dimethyl-1,3-butadiene, 98%, stab. with 100ppm BHT
Description
2,3-Dimethyl-1,3-butadiene undergoes Diels Alder cycloaddition reaction and it also participates in polymerization reactions. It is also involved in manufacturing of synthetic rubber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3-Dimethyl-1,3-butadiene undergoes Diels Alder cycloaddition reaction and it also participates in polymerization reactions. It is also involved in manufacturing of synthetic rubber.
Solubility
Miscible with chloroform. Immiscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
2,3-Dimethyl-1,3-butadiene undergoes Diels Alder cycloaddition reaction and it also participates in polymerization reactions. It is also involved in manufacturing of synthetic rubber.
Solubility
Miscible with chloroform. Immiscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | -76°C |
| Density | 0.726 |
| Boiling Point | 68°C to 69°C |
| Flash Point | −1°C (30°F) |
| Refractive Index | 1.438 |
| Quantity | 10 g |
| UN Number | UN3295 |
| Beilstein | 605285 |
| Merck Index | 14,3238 |
| Solubility Information | Miscible with chloroform. Immiscible with water. |
| Show More |
Safety and Handling
| Hazard Category | H225 |
| Hazard Statement | GHS H Statement H225-H304 Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S. |
| EINECSNumber | 208-172-0 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only