accessibility menu, dialog, popup

UserName

H3CCH2CH3H2C

CAS RN 513-81-5

IUPAC Name: 2,3-dimethylbuta-1,3-diene
Synonyms: dimethylbutadiene
Molecular Weight (g/mol): 82.15
Molecular Formula: C6H10
InChi Key: SDJHPPZKZZWAKF-UHFFFAOYSA-N
SMILES: CC(=C)C(C)=C
Boiling Point 
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)