missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-1,3-butadiene, 98%, stab. with 100ppm BHT

Catalog No. AAL0420709
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0420709 10 g
AAL0420718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0420709 Supplier Thermo Scientific Chemicals Supplier No. L0420709

CAS: 513-81-5 | C6H10 | 82.15 g/mol

2,3-Dimethyl-1,3-butadiene undergoes Diels Alder cycloaddition reaction and it also participates in polymerization reactions. It is also involved in manufacturing of synthetic rubber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dimethyl-1,3-butadiene undergoes Diels Alder cycloaddition reaction and it also participates in polymerization reactions. It is also involved in manufacturing of synthetic rubber.

Solubility
Miscible with chloroform. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 513-81-5
Molecular Formula C6H10
Molecular Weight (g/mol) 82.15
MDL Number MFCD00008595
InChI Key SDJHPPZKZZWAKF-UHFFFAOYSA-N
Synonym 2,3-dimethyl-1,3-butadiene, biisopropenyl, 2,3-dimethylbutadiene, diisopropenyl, 2,3-dimethylenebutane, 1,3-butadiene, 2,3-dimethyl, unii-61tuu25hco, 61tuu25hco, 2,3-dimethyl-buta-1,3-diene, 2,3-diene
PubChem CID 10566
IUPAC Name 2,3-dimethylbuta-1,3-diene
SMILES CC(=C)C(C)=C

Specifications

Melting Point -76°C
Density 0.726
Boiling Point 68°C to 69°C
Flash Point −1°C (30°F)
Refractive Index 1.438
Quantity 10 g
UN Number UN3295
Beilstein 605285
Merck Index 14,3238
Solubility Information Miscible with chloroform. Immiscible with water.
Formula Weight 82.15
Percent Purity 98%
Chemical Name or Material 2,3-Dimethyl-1,3-butadiene, stabilized with 100ppm BHT
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 208-172-0
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only