missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxytoluene, 98+%

Catalog No. AAA1161714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1161714 25 g
AAA1161706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1161714 Supplier Thermo Scientific Chemicals Supplier No. A1161714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4463-33-6 | C9H12O2 | 152.193 g/mol

2,3-Dimethoxytoluene has been used to prepare 6,7-dimethoxy-4-hydroxyisochroman-3-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dimethoxytoluene has been used to prepare 6,7-dimethoxy-4-hydroxyisochroman-3-one.

Solubility
Soluble in Dichloromethane. Not miscible in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.

Chemical Identifiers

CAS 4463-33-6
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00008379
InChI Key WMXFNCKPYCAIQW-UHFFFAOYSA-N
Synonym 2,3-dimethoxytoluene, 3-methylveratrole, benzene, 1,2-dimethoxy-3-methyl, dimethoxytoluene, 1,2-dimethoxy-3-methyl-benzene, acmc-1ae12, 1-methyl-2,3-dimethoxybenzene, 3-methylcatechol, dimethyl ether, #
PubChem CID 78215
IUPAC Name 1,2-dimethoxy-3-methylbenzene
SMILES CC1=C(C(=CC=C1)OC)OC

Specifications

Density 1.024
Boiling Point 202°C to 203°C
Flash Point 85°C (185°F)
Odor Aromatic
Refractive Index 1.514
Quantity 25 g
Beilstein 1940962
Solubility Information Soluble in Dichloromethane. Not miscible in water.
Formula Weight 152.19
Percent Purity ≥98%
Chemical Name or Material 2,3-Dimethoxytoluene
Show More Show Less
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 224-726-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only