missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxypyridine

Catalog No. AAL1962414
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1962414 25 g
AAL1962403 1 g
AAL1962406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1962414 Supplier Thermo Scientific Chemicals Supplier No. L1962414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 52605-97-7 | C7H9NO2 | 139.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52605-97-7
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD03095043
InChI Key QHUHPERZCBUMRK-UHFFFAOYSA-N
Synonym pyridine, 2,3-dimethoxy, dimethoxypyridine, pubchem2412, zlchem 1174, 2,3-dimethoxy-pyridine, acmc-1argu, tpc-py047
PubChem CID 4657809
IUPAC Name 2,3-dimethoxypyridine
SMILES COC1=C(N=CC=C1)OC

Specifications

Density 1.104
Boiling Point 98°C to 100°C (16 mmHg)
Refractive Index 1.5205
Quantity 25 g
Beilstein 1526067
Formula Weight 139.15
Chemical Name or Material 2,3-Dimethoxypyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only