accessibility menu, dialog, popup

UserName

H3CONOCH3

CAS RN 52605-97-7

IUPAC Name: 2,3-dimethoxypyridine
Synonyms: 2,3-dimethoxypyridine
Molecular Weight (g/mol): 139.15
Molecular Formula: C7H9NO2
InChi Key: QHUHPERZCBUMRK-UHFFFAOYSA-N
SMILES: COC1=CC=CN=C1OC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)