missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxycinnamic acid, predominantly trans, 98+%

Catalog No. AAA1593609
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1593609 10 g
AAA1593618 50 g
AAA1593630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1593609 Supplier Thermo Scientific Chemicals Supplier No. A1593609
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7345-82-6 | C11H12O4 | 208.213 g/mol

2,3-Dimethoxycinnamic acid is used in the synthesis of (2E,2E,2E)-N,N,N-(nitrilotri-2,1-ethanediyl)tris[3-(2,3-dimethoxyphenyl)-2-propenamide].

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dimethoxycinnamic acid is used in the synthesis of (2E,2′E,2′′E)-N,N′,N′′-(nitrilotri-2,1-ethanediyl)tris[3-(2,3-dimethoxyphenyl)-2-propenamide].

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 7345-82-6
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD00004376
InChI Key QAXPUWGAGVERSJ-VOTSOKGWSA-N
Synonym 2,3-dimethoxycinnamic acid, trans-2,3-dimethoxycinnamic acid, 2e-3-2,3-dimethoxyphenyl acrylic acid, e-3-2,3-dimethoxyphenyl acrylic acid, 2,3-dimethoxy cinnamic acid, 3-2,3-dimethoxyphenyl acrylic acid, 2e-3-2,3-dimethoxyphenyl prop-2-enoic acid, 2-propenoic acid, 3-2,3-dimethoxyphenyl, e-3-2,3-dimethoxyphenyl prop-2-enoic acid, e-3-2,3-dimethoxy-phenyl-acrylic acid
PubChem CID 735842
IUPAC Name (E)-3-(2,3-dimethoxyphenyl)prop-2-enoic acid
SMILES COC1=CC=CC(=C1OC)C=CC(=O)O

Specifications

Melting Point 179°C to 183°C
Quantity 10 g
Beilstein 2695122
Formula Weight 208.21
Percent Purity ≥98%
Chemical Name or Material 2,3-Dimethoxycinnamic acid, predominantly trans
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 231-249-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only