missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxybenzoic Acid 98.0+%, TCI America™

Catalog No. p-7126331
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
D063025G 25 g
D0630500G 500 g
2 options

Catalog No. D063025G

Supplier: TCI America D063025G

Only null left

Chemical Identifiers

CAS 1521-38-6
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00002432
InChI Key FODBVCSYJKNBLO-UHFFFAOYSA-N
Synonym o-veratric acid, benzoic acid, 2,3-dimethoxy, benzoic acid, dimethoxy, 2,3-dimethoxy benzoic acid, dimethoxybenzoic acid,2,3, o-veratric acid 6ci,7ci,8ci, 2,3-dimethoxybenzoicacid, dimethoxybenzoic acid, pubchem4948, 2,3-dimethoxy-benzoicaci
PubChem CID 15204
IUPAC Name 2,3-dimethoxybenzoic acid
SMILES COC1=CC=CC(C(O)=O)=C1OC

Specifications

Melting Point 124°C
Color White
Quantity 25 g
Formula Weight 182.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dimethoxybenzoic Acid
EINECSNumber (3)-1645
RTECSNumber DG8598700
TSCA No