missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxybenzoic acid, 99%

Catalog No. AAA1459209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1459209 10 g
AAA1459218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1459209 Supplier Thermo Scientific Chemicals Supplier No. A1459209
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1521-38-6 | C9H10O4 | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1521-38-6
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00002432
InChI Key FODBVCSYJKNBLO-UHFFFAOYSA-N
Synonym o-veratric acid, benzoic acid, 2,3-dimethoxy, benzoic acid, dimethoxy, 2,3-dimethoxy benzoic acid, dimethoxybenzoic acid,2,3, o-veratric acid 6ci,7ci,8ci, 2,3-dimethoxybenzoicacid, dimethoxybenzoic acid, pubchem4948, 2,3-dimethoxy-benzoicaci
PubChem CID 15204
IUPAC Name 2,3-dimethoxybenzoic acid
SMILES COC1=CC=CC(C(O)=O)=C1OC

Specifications

Melting Point 121°C to 124°C
Odor Odorless
Quantity 10 g
Beilstein 2210858
Formula Weight 182.18
Percent Purity 99%
Chemical Name or Material 2,3-Dimethoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-188-4
RTECSNumber DG8598700
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only