missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxyaniline 98.0+%, TCI America™

Catalog No. p-7127212
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22841G 1 g
D22845G 5 g
D228425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D22841G Supplier TCI America Supplier No. D22841G

Chemical Identifiers

CAS 6299-67-8
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.181
MDL Number MFCD00180769
InChI Key HEZIOZBMPKPOER-UHFFFAOYSA-N
PubChem CID 239603
IUPAC Name 2,3-dimethoxyaniline
SMILES COC1=CC=CC(=C1OC)N

Specifications

Boiling Point 93°C
Quantity 1 g
Formula Weight 153.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,3-Dimethoxyaniline
EINECSNumber (3)-0616
TSCA No
Recommended Storage Refrigerator