missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate, 95%

Catalog No. AAH56578MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH56578MD 250 mg
AAH5657803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH56578MD Supplier Thermo Scientific Chemicals Supplier No. H56578MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 207233-91-8 | C3H7O3S3 | 187.27 g/mol

Sodium 2,3-dimercaptopropanesulfonate monohydrate has been used in a study to assess its use as an antidote.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 2,3-dimercaptopropanesulfonate monohydrate has been used in a study to assess its use as an antidote.

Solubility
Soluble in Water: 100 mg/ml

Notes
Store at 4°C. Incompatible with oxidizing agents. Keep container tightly closed.

Chemical Identifiers

CAS 207233-91-8
Molecular Formula C3H7O3S3
Molecular Weight (g/mol) 187.27
MDL Number MFCD00149543
InChI Key JLVSRWOIZZXQAD-GSVOUGTGSA-M
Synonym sodium 2,3-dimercaptopropanesulfonate monohydrate, unithiol, dl-2,3-dimercapto-1-propanesulfonic acid sodium salt monohydrate, sodium 2,3-dimercaptopropane-1-sulfonate hydrate, 2,3-dimercapto-1-propanesulfonic acid sodium salt monohydrate, sodium 2,3-dimercaptopropanesulfonate hydrate, dmps, sodium 2,3-disulfanylpropane-1-sulfonate hydrate, unitiol monohydrate
PubChem CID 23676755
SMILES [O-]S(=O)(=O)C[C@H](S)CS

Specifications

Melting Point ∽229°C (decomposition)
Quantity 250 mg
Beilstein 3734863
Merck Index 14,3210
Solubility Information Soluble in Water: 100mg/ml
IUPAC Name (2R)-2,3-disulfanylpropane-1-sulfonate
Formula Weight 228.30 (210.28 Anhydrous)
Percent Purity 98%
Chemical Name or Material 2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate
EINECSNumber 223-796-3
RTECSNumber TZ6420000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only