CAS RN 207233-91-8
IUPAC Name:
(2R)-2,3-disulfanylpropane-1-sulfonate
Synonyms:
(2R)-2,3-disulfanylpropane-1-sulfonate
Molecular Weight (g/mol):
187.27
Molecular Formula:
C3H7O3S3
InChi Key:
JLVSRWOIZZXQAD-GSVOUGTGSA-M
SMILES:
[O-]S(=O)(=O)C[C@H](S)CS
1
–
1
of
1
results
1
Promotions Available
2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate, 95%
2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate, 95%
CAS: 207233-91-8 Molecular Formula: C3H7O3S3 Molecular Weight (g/mol): 187.27 MDL Number: MFCD00149543 InChI Key: JLVSRWOIZZXQAD-GSVOUGTGSA-M Synonym: sodium 2,3-dimercaptopropanesulfonate monohydrate,unithiol,dl-2,3-dimercapto-1-propanesulfonic acid sodium salt monohydrate,sodium 2,3-dimercaptopropane-1-sulfonate hydrate,2,3-dimercapto-1-propanesulfonic acid sodium salt monohydrate,sodium 2,3-dimercaptopropanesulfonate hydrate,dmps,sodium 2,3-disulfanylpropane-1-sulfonate hydrate,unitiol monohydrate PubChem CID: 23676755 SMILES: [O-]S(=O)(=O)C[C@H](S)CS