missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydroxyquinoxaline, 98%

Catalog No. p-7044644
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0758114 25 g
AAL0758122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0758114 Supplier Thermo Scientific Chemicals Supplier No. L0758114
Only null left

CAS: 15804-19-0 | C8H6N2O2 | 162.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15804-19-0
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00006723
InChI Key ABJFBJGGLJVMAQ-UHFFFAOYSA-N
Synonym 2,3-dihydroxyquinoxaline, quinoxaline-2,3-diol, quinoxaline-2,3 1h,4h-dione, 2,3-quinoxalinediol, 1,4-dihydro-2,3-quinoxalinedione, 2,3-quinoxalinedione, 1,4-dihydro, usaf ek-6232, 2,3 1h,4h-quinoxalinedione, quinoxaline, 2,3-dihydroxy, unii-apo55iz9e1
PubChem CID 27491
IUPAC Name 1,4-dihydroquinoxaline-2,3-dione
SMILES C1=CC=C2C(=C1)NC(=O)C(=O)N2

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 136884
Formula Weight 162.15
Percent Purity 98%
Chemical Name or Material 2,3-Dihydroxyquinoxaline
EINECSNumber 239-901-0
RTECSNumber VD2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only