accessibility menu, dialog, popup

UserName

OHNNHO

CAS RN 15804-19-0

IUPAC Name: 1,2,3,4-tetrahydroquinoxaline-2,3-dione
Synonyms: quinoxalinedione
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: ABJFBJGGLJVMAQ-UHFFFAOYSA-N
SMILES: O=C1NC2=CC=CC=C2NC1=O
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)