missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydroxynaphthalene, 98%

Catalog No. p-7081363
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1130718 50 g
AAA1130730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1130718 Supplier Thermo Scientific Chemicals Supplier No. A1130718

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 92-44-4 | C10H8O2 | 160.17 g/mol

2,3-Dihydroxynaphthalene is used in making dyes, pigments, fluorescent whiteners, tanning agents, antioxidants, and antiseptics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dihydroxynaphthalene is used in making dyes, pigments, fluorescent whiteners, tanning agents, antioxidants, and antiseptics.

Solubility
Slightly soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 92-44-4
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
MDL Number MFCD00004073
InChI Key JRNGUTKWMSBIBF-UHFFFAOYSA-N
Synonym 2,3-dihydroxynaphthalene, 2,3-naphthalenediol, 2,3-dihydroxynapthalene, 2,3-dihydroxy naphthalene, 2,3-dihydroxynaphthlene, naphthalenediol-2,3, 3-hydroxy-2-naphthol, 2,3dihydroxynaphthalene, acmc-209rfr, 2,3-dihydroxylnaphthalene
PubChem CID 7091
ChEBI CHEBI:38135
IUPAC Name naphthalene-2,3-diol
SMILES OC1=C(O)C=C2C=CC=CC2=C1

Specifications

Melting Point 162°C to 165°C
Flash Point 175°C (347°F)
Quantity 50 g
Beilstein 742375
Solubility Information Slightly soluble in water
Formula Weight 160.17
Percent Purity 98%
Chemical Name or Material 2,3-Dihydroxynaphthalene
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
EINECSNumber 202-156-7
RTECSNumber QJ4750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only