missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydroxynaphthalene, 97%

Catalog No. p-112065
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC115020500 50 g Plastic bottle
AC115022500 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC115020500 Supplier Thermo Scientific Chemicals Supplier No. 115020500
Only null left

CAS: 92-44-4 | C10H8O2 | 160.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 92-44-4
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
MDL Number MFCD00004073
InChI Key JRNGUTKWMSBIBF-UHFFFAOYSA-N
Synonym 2,3-dihydroxynaphthalene, 2,3-naphthalenediol, 2,3-dihydroxynapthalene, 2,3-dihydroxy naphthalene, 2,3-dihydroxynaphthlene, naphthalenediol-2,3, 3-hydroxy-2-naphthol, 2,3dihydroxynaphthalene, acmc-209rfr, 2,3-dihydroxylnaphthalene
PubChem CID 7091
ChEBI CHEBI:38135
IUPAC Name naphthalene-2,3-diol
SMILES OC1=C(O)C=C2C=CC=CC2=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 162.0°C to 166.0°C
Color Beige-Gray to White
Flash Point 175°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula C10H6(OH)2
Quantity 50 g
Beilstein 06, 982
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 160.17
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2, 3-Dihydroxynaphthalene, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-156-7
RTECSNumber QJ4750000
TSCA TSCA

RUO – Research Use Only