missing translation for 'onlineSavingsMsg'
Learn More

2',3'-Dihydroxy-4'-methoxyacetophenone Hydrate 98.0+%, TCI America™

Catalog No. D21875G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21875G 5 g
D218725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21875G Supplier TCI America Supplier No. D21875G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 708-53-2
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00180772
InChI Key VCONERRCKOKCHE-UHFFFAOYSA-N
PubChem CID 12820
IUPAC Name 1-(2,3-dihydroxy-4-methoxyphenyl)ethanone
SMILES CC(=O)C1=C(C(=C(C=C1)OC)O)O

Specifications

Melting Point 133°C
Quantity 5 g
Synonym Gallacetophenone-4′C-methyl Ether
Formula Weight 182.18
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2′,3′-Dihydroxy-4′-methoxyacetophenone Hydrate