accessibility menu, dialog, popup

UserName

H3COCH3OHOHO

CAS RN 708-53-2

IUPAC Name: 1-(2,3-dihydroxy-4-methoxyphenyl)ethan-1-one
Synonyms: 1-(2,3-dihydroxy-4-methoxyphenyl)ethanone
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: VCONERRCKOKCHE-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(C)=O)C(O)=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results