missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dihydrobenzofuran-7-carboxylic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 172°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 164.16 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,3-Dihydrobenzofuran-7-carboxylic Acid |
Safety and Handling
| TSCA | No |