accessibility menu, dialog, popup

UserName

CAS RN 35700-40-4

IUPAC Name: 2,3-dihydro-1-benzofuran-7-carboxylic acid
Synonyms: 2,3-dihydro-1-benzofuran-7-carboxylic acid
Molecular Weight (g/mol): 164.16
Molecular Formula: C9H8O3
InChi Key: UHXBMSNEECJPSX-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2OCCC2=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)