missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrobenzofuran-7-carboxylic Acid 98.0+%, TCI America™

Catalog No. d24461g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
D24461G 1 g
D24465G 5 g
2 options

Catalog No. D24461G

Supplier: TCI America D24461G

Only null left

Chemical Identifiers

CAS 35700-40-4
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00671526
InChI Key UHXBMSNEECJPSX-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-7-carboxylic acid, 2,3-dihydrobenzo b furan-7-carboxylic acid, 7-coumarancarboxylic acid, 2,3-dihydro-7-benzofurancarboxylic acid, 7-benzofurancarboxylicacid, 2,3-dihydro, 7-benzofurancarboxylic acid, 2,3-dihydro, pubchem16734, acmc-209iiq, dihydrobenzofurancarboxylicacid, 7-dihydrobenzofuran carboxylic acid
PubChem CID 2795014
IUPAC Name 2,3-dihydro-1-benzofuran-7-carboxylic acid
SMILES C1COC2=C(C=CC=C21)C(=O)O

Specifications

Melting Point 172°C
Color White-Yellow
Quantity 1 g
Formula Weight 164.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dihydrobenzofuran-7-carboxylic Acid