missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrobenzofuran-5-carboxaldehyde 96.0+%, TCI America™

Catalog No. p-7127956
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38575G 5 g
D385725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38575G Supplier TCI America Supplier No. D38575G

Chemical Identifiers

CAS 55745-70-5
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
MDL Number MFCD00068058
InChI Key WEBVDBDZSOJGPB-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-5-carboxaldehyde, 2,3-dihydrobenzo b furan-5-carbaldehyde, 2,3-dihydrobenzofuran-5-carbaldehyde, 2,3-dihydrobenzo b furan-5-carboxaldehyde, 2,3-dihyro-1-benzofuran-5-carbaldehyde, 2,3-dihydro-benzofuran-5-carbaldehyde, 5-coumarancarboxaldehyde, 2,3-dihydro-5-benzofurancarboxaldehyde, 5-benzofurancarboxaldehyde, 2,3-dihydro, 2,3-dihydro-5-benzofurancarbaldehyde
PubChem CID 735901
IUPAC Name 2,3-dihydro-1-benzofuran-5-carbaldehyde
SMILES O=CC1=CC=C2OCCC2=C1

Specifications

Color Yellow
Boiling Point 96°C
Quantity 5 g
Formula Weight 148.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dihydrobenzofuran-5-carboxaldehyde