accessibility menu, dialog, popup

UserName

CAS RN 55745-70-5

IUPAC Name: 2,3-dihydro-1-benzofuran-5-carbaldehyde
Synonyms: 2,3-dihydro-1-benzofuran-5-carbaldehyde
Molecular Weight (g/mol): 148.16
Molecular Formula: C9H8O2
InChi Key: WEBVDBDZSOJGPB-UHFFFAOYSA-N
SMILES: O=CC1=CC=C2OCCC2=C1
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)