missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrobenzo[b]furan-5-ylmethylamine hydrochloride, 97%, Thermo Scientific™

Chemical Identifiers

CAS 55745-74-9
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00728869
InChI Key WQXWNTPLZFVZNX-UHFFFAOYSA-N
Synonym 5-aminomethyl-2,3-dihydrobenzo b furan, 2,3-dihydrobenzofuran-5-yl methanamine, 2,3-dihydrobenzo b furan-5-ylmethylamine, 1-2,3-dihydro-1-benzofuran-5-yl methanamine, 2,3-dihydro-benzofuran-5-ylmethylamine, 2,3-dihydro-1-benzofuran-5-yl methanamine, pubchem6985, 2,3-dihydrobenzofuran-5-ylmethanamine, c-2,3-dihydrobenzofuran-5-ylmethylamine
PubChem CID 2735367
IUPAC Name 2,3-dihydro-1-benzofuran-5-ylmethanamine
SMILES NCC1=CC2=C(OCC2)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC00813CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC00813DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

55745-74-9
149.19
WQXWNTPLZFVZNX-UHFFFAOYSA-N
2735367
NCC1=CC2=C(OCC2)C=C1
C9H11NO
MFCD00728869
5-aminomethyl-2,3-dihydrobenzo b furan, 2,3-dihydrobenzofuran-5-yl methanamine, 2,3-dihydrobenzo b furan-5-ylmethylamine, 1-2,3-dihydro-1-benzofuran-5-yl methanamine, 2,3-dihydro-benzofuran-5-ylmethylamine, 2,3-dihydro-1-benzofuran-5-yl methanamine, pubchem6985, 2,3-dihydrobenzofuran-5-ylmethanamine, c-2,3-dihydrobenzofuran-5-ylmethylamine
2,3-dihydro-1-benzofuran-5-ylmethanamine

Specifications

55745-74-9
100.0
C9H11NO
5-aminomethyl-2,3-dihydrobenzo b furan, 2,3-dihydrobenzofuran-5-yl methanamine, 2,3-dihydrobenzo b furan-5-ylmethylamine, 1-2,3-dihydro-1-benzofuran-5-yl methanamine, 2,3-dihydro-benzofuran-5-ylmethylamine, 2,3-dihydro-1-benzofuran-5-yl methanamine, pubchem6985, 2,3-dihydrobenzofuran-5-ylmethanamine, c-2,3-dihydrobenzofuran-5-ylmethylamine
NCC1=CC2=C(OCC2)C=C1
149.19
185.65
2, 3-Dihydrobenzo[b]furan-5-ylmethylamine hydrochloride
96.5
97%
MFCD00728869
WQXWNTPLZFVZNX-UHFFFAOYSA-N
2,3-dihydro-1-benzofuran-5-ylmethanamine
2735367
97%
Videos
SDS
Documents

Documents