missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dihydrobenzo[b]furan-5-ylmethylamine hydrochloride, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC00813CB
| Quantity | 250mg |
|---|---|
| Packaging | Amber Glass Bottle |
Chemical Identifiers
| 55745-74-9 | |
| 149.19 | |
| WQXWNTPLZFVZNX-UHFFFAOYSA-N | |
| 2735367 | |
| NCC1=CC2=C(OCC2)C=C1 |
| C9H11NO | |
| MFCD00728869 | |
| 5-aminomethyl-2,3-dihydrobenzo b furan, 2,3-dihydrobenzofuran-5-yl methanamine, 2,3-dihydrobenzo b furan-5-ylmethylamine, 1-2,3-dihydro-1-benzofuran-5-yl methanamine, 2,3-dihydro-benzofuran-5-ylmethylamine, 2,3-dihydro-1-benzofuran-5-yl methanamine, pubchem6985, 2,3-dihydrobenzofuran-5-ylmethanamine, c-2,3-dihydrobenzofuran-5-ylmethylamine | |
| 2,3-dihydro-1-benzofuran-5-ylmethanamine |
Specifications
| 2, 3-Dihydrobenzo[b]furan-5-ylmethylamine hydrochloride | |
| 96.5 | |
| 97% | |
| C9H11NO | |
| 250mg | |
| WQXWNTPLZFVZNX-UHFFFAOYSA-N | |
| 2,3-dihydro-1-benzofuran-5-ylmethanamine | |
| 2735367 | |
| 97% |
| 55745-74-9 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD00728869 | |
| 5-aminomethyl-2,3-dihydrobenzo b furan, 2,3-dihydrobenzofuran-5-yl methanamine, 2,3-dihydrobenzo b furan-5-ylmethylamine, 1-2,3-dihydro-1-benzofuran-5-yl methanamine, 2,3-dihydro-benzofuran-5-ylmethylamine, 2,3-dihydro-1-benzofuran-5-yl methanamine, pubchem6985, 2,3-dihydrobenzofuran-5-ylmethanamine, c-2,3-dihydrobenzofuran-5-ylmethylamine | |
| NCC1=CC2=C(OCC2)C=C1 | |
| 149.19 | |
| 185.65 |