missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrobenzo[b]furan-5-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 76429-73-7
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
InChI Key YXYOLVAXVPOIMA-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-5-carboxylic acid, 2,3-dihydrobenzo b furan-5-carboxylic acid, 5-benzofurancarboxylic acid, 2,3-dihydro, pubchem9983, pubchem14824, maybridge1_002497, ksc377a5f, dihydrobenzofuran-5-carboxylic acid, 5-carboxy-2,3-dihydrobenzo b furan
PubChem CID 676583
IUPAC Name 2,3-dihydro-1-benzofuran-5-carboxylic acid
SMILES C1COC2=C1C=C(C=C2)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC00801EA
View Documents
N/A 10g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

76429-73-7
164.16
2,3-dihydrobenzofuran-5-carboxylic acid, 2,3-dihydrobenzo b furan-5-carboxylic acid, 5-benzofurancarboxylic acid, 2,3-dihydro, pubchem9983, pubchem14824, maybridge1_002497, ksc377a5f, dihydrobenzofuran-5-carboxylic acid, 5-carboxy-2,3-dihydrobenzo b furan
2,3-dihydro-1-benzofuran-5-carboxylic acid
C9H8O3
YXYOLVAXVPOIMA-UHFFFAOYSA-N
676583
C1COC2=C1C=C(C=C2)C(=O)O

Specifications

76429-73-7
C9H8O3
YXYOLVAXVPOIMA-UHFFFAOYSA-N
2,3-dihydro-1-benzofuran-5-carboxylic acid
676583
97%
97%
2,3-dihydrobenzofuran-5-carboxylic acid, 2,3-dihydrobenzo b furan-5-carboxylic acid, 5-benzofurancarboxylic acid, 2,3-dihydro, pubchem9983, pubchem14824, maybridge1_002497, ksc377a5f, dihydrobenzofuran-5-carboxylic acid, 5-carboxy-2,3-dihydrobenzo b furan
C1COC2=C1C=C(C=C2)C(=O)O
164.16
164.16
2, 3-Dihydrobenzo[b]furan-5-carboxylic acid
Videos
SDS
Documents

Documents