CAS RN 76429-73-7
IUPAC Name:
2,3-dihydro-1-benzofuran-5-carboxylic acid
Synonyms:
2,3-dihydro-1-benzofuran-5-carboxylic acid
Molecular Weight (g/mol):
164.16
Molecular Formula:
C9H8O3
InChi Key:
YXYOLVAXVPOIMA-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC2=C(OCC2)C=C1
1
–
1
of
1
results
1
2,3-Dihydrobenzo[b]furan-5-carboxylic acid, 97%, Thermo Scientific™
CAS: 76429-73-7 Molecular Formula: C9H8O3 Molecular Weight (g/mol): 164.16 InChI Key: YXYOLVAXVPOIMA-UHFFFAOYSA-N Synonym: 2,3-dihydrobenzofuran-5-carboxylic acid,2,3-dihydrobenzo b furan-5-carboxylic acid,5-benzofurancarboxylic acid, 2,3-dihydro,pubchem9983,pubchem14824,maybridge1_002497,ksc377a5f,dihydrobenzofuran-5-carboxylic acid,5-carboxy-2,3-dihydrobenzo b furan PubChem CID: 676583 IUPAC Name: 2,3-dihydro-1-benzofuran-5-carboxylic acid SMILES: C1COC2=C1C=C(C=C2)C(=O)O