missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrobenzo[b]furan-5-carboxaldehyde, 97%

Catalog No. p-7040022
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5960203 1 g
AAH5960206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5960203 Supplier Thermo Scientific Chemicals Supplier No. H5960203
Only null left

CAS: 55745-70-5 | C9H8O2 | 148.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55745-70-5
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
MDL Number MFCD00068058
InChI Key WEBVDBDZSOJGPB-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-5-carboxaldehyde, 2,3-dihydrobenzo b furan-5-carbaldehyde, 2,3-dihydrobenzofuran-5-carbaldehyde, 2,3-dihydrobenzo b furan-5-carboxaldehyde, 2,3-dihyro-1-benzofuran-5-carbaldehyde, 2,3-dihydro-benzofuran-5-carbaldehyde, 5-coumarancarboxaldehyde, 2,3-dihydro-5-benzofurancarboxaldehyde, 5-benzofurancarboxaldehyde, 2,3-dihydro, 2,3-dihydro-5-benzofurancarbaldehyde
PubChem CID 735901
IUPAC Name 2,3-dihydro-1-benzofuran-5-carbaldehyde
SMILES O=CC1=CC=C2OCCC2=C1

Specifications

Density 1.177
Boiling Point 88°C to 89°C (0.1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.6
Quantity 1 g
Beilstein 124062
Formula Weight 148.16
Percent Purity 97%
Chemical Name or Material 2,3-Dihydrobenzo[b]furan-5-carboxaldehyde
Hazard Category H334
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P285-P304+P341-P342+P311-P501c
EINECSNumber 259-788-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only