missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1H-benz[de]isoquinoline, 97%

CAS: 22817-26-1 | C12H11N | 169.227 g/mol

Supplier:  Thermo Scientific Chemicals L1561303

 View more versions of this product

Catalog No. AAL1561303


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,3-Dihydro-1H-benz[de]isoquinoline
96°C to 101°C
C12H11N
1 g
134303
NBWCGDRKSOXZNI-UHFFFAOYSA-N
2,3-dihydro-1H-benzo[de]isoquinoline
208526
97%
22817-26-1
120°C (0.5 mmHg)
MFCD01075706
UN3259
2,3-dihydro-1h-benzo de isoquinoline, 2,3-dihydro-1h-benz de isoquinoline, 1h-benz de isoquinoline,2,3-dihydro, pubchem14095, acmc-20a5ts, 2,3-dihydro-1 h-benz de isochinolin, 3-azatricyclo 7.3.1.0?,__ trideca-1 13 ,5,7,9,11-pentaene, 3-azatricyclo 7.3.1.0?, 1 3 trideca-1 12 ,5,7,9 13 ,10-pentaene, 3-azatricyclo 7.3.1.0?, 1 3 trideca-1 13 ,5,7,9,11-pentaene
C1C2=CC=CC3=C2C(=CC=C3)CN1
169.227
169.23

Chemical Identifiers

22817-26-1
169.227
NBWCGDRKSOXZNI-UHFFFAOYSA-N
208526
C1C2=CC=CC3=C2C(=CC=C3)CN1
C12H11N
MFCD01075706
2,3-dihydro-1h-benzo de isoquinoline, 2,3-dihydro-1h-benz de isoquinoline, 1h-benz de isoquinoline,2,3-dihydro, pubchem14095, acmc-20a5ts, 2,3-dihydro-1 h-benz de isochinolin, 3-azatricyclo 7.3.1.0?,__ trideca-1 13 ,5,7,9,11-pentaene, 3-azatricyclo 7.3.1.0?, 1 3 trideca-1 12 ,5,7,9 13 ,10-pentaene, 3-azatricyclo 7.3.1.0?, 1 3 trideca-1 13 ,5,7,9,11-pentaene
2,3-dihydro-1H-benzo[de]isoquinoline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

TSCA : No

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only