missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1H-benz[de]isoquinoline, 97%

Catalog No. p-7046662
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL15613MD 250 mg
AAL1561303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL15613MD Supplier Thermo Scientific Chemicals Supplier No. L15613MD

CAS: 22817-26-1 | C12H11N | 169.227 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22817-26-1
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.227
MDL Number MFCD01075706
InChI Key NBWCGDRKSOXZNI-UHFFFAOYSA-N
Synonym 2,3-dihydro-1h-benzo de isoquinoline, 2,3-dihydro-1h-benz de isoquinoline, 1h-benz de isoquinoline,2,3-dihydro, pubchem14095, acmc-20a5ts, 2,3-dihydro-1 h-benz de isochinolin, 3-azatricyclo 7.3.1.0?,__ trideca-1 13 ,5,7,9,11-pentaene, 3-azatricyclo 7.3.1.0?, 1 3 trideca-1 12 ,5,7,9 13 ,10-pentaene, 3-azatricyclo 7.3.1.0?, 1 3 trideca-1 13 ,5,7,9,11-pentaene
PubChem CID 208526
IUPAC Name 2,3-dihydro-1H-benzo[de]isoquinoline
SMILES C1C2=CC=CC3=C2C(=CC=C3)CN1

Specifications

Melting Point 96°C to 101°C
Boiling Point 120°C (0.5 mmHg)
Quantity 250 mg
UN Number UN3259
Beilstein 134303
Formula Weight 169.23
Percent Purity 97%
Chemical Name or Material 2,3-Dihydro-1H-benz[de]isoquinoline
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No

RUO – Research Use Only