missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dihydro-1,4-benzodioxin-5-ylmethylamine, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 261633-71-0 |
|---|---|
| Molecular Formula | C9H11NO2 |
| Molecular Weight (g/mol) | 165.192 |
| InChI Key | OKRXRKUGZVMPKX-UHFFFAOYSA-N |
| Synonym | 2,3-dihydrobenzo b 1,4 dioxin-5-yl methanamine, c-2,3-dihydro-benzo 1,4 dioxin-5-yl-methylamine, 1,4-benzodioxin-5-methanamine, 2,3-dihydro, 2,3-dihydro-1,4-benzodioxin-5-ylmethylamine, 5-aminomethyl-1,4-benzodioxane, 1-2,3-dihydro-1,4-benzodioxin-5-yl methanamine, c-2,3-dihydrobenzo 1,4 dioxin-5-yl methylamine |
| PubChem CID | 2795040 |
| IUPAC Name | 2,3-dihydro-1,4-benzodioxin-5-ylmethanamine |
| SMILES | C1COC2=C(C=CC=C2O1)CN |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
CC01413CB
|
N/A | 250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC01413DA
|
N/A | 1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC01413DE
|
N/A | 5g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 261633-71-0 | |
| 165.192 | |
| 2,3-dihydrobenzo b 1,4 dioxin-5-yl methanamine, c-2,3-dihydro-benzo 1,4 dioxin-5-yl-methylamine, 1,4-benzodioxin-5-methanamine, 2,3-dihydro, 2,3-dihydro-1,4-benzodioxin-5-ylmethylamine, 5-aminomethyl-1,4-benzodioxane, 1-2,3-dihydro-1,4-benzodioxin-5-yl methanamine, c-2,3-dihydrobenzo 1,4 dioxin-5-yl methylamine | |
| 2,3-dihydro-1,4-benzodioxin-5-ylmethanamine |
| C9H11NO2 | |
| OKRXRKUGZVMPKX-UHFFFAOYSA-N | |
| 2795040 | |
| C1COC2=C(C=CC=C2O1)CN |
Specifications
| 261633-71-0 | |
| C9H11NO2 | |
| OKRXRKUGZVMPKX-UHFFFAOYSA-N | |
| 2,3-dihydro-1,4-benzodioxin-5-ylmethanamine | |
| 2795040 | |
| 95% |
| 95% | |
| 2,3-dihydrobenzo b 1,4 dioxin-5-yl methanamine, c-2,3-dihydro-benzo 1,4 dioxin-5-yl-methylamine, 1,4-benzodioxin-5-methanamine, 2,3-dihydro, 2,3-dihydro-1,4-benzodioxin-5-ylmethylamine, 5-aminomethyl-1,4-benzodioxane, 1-2,3-dihydro-1,4-benzodioxin-5-yl methanamine, c-2,3-dihydrobenzo 1,4 dioxin-5-yl methylamine | |
| C1COC2=C(C=CC=C2O1)CN | |
| 165.192 | |
| 201.65 | |
| 2, 3-Dihydro-1, 4-benzodioxin-5-ylmethylamine hydrochloride |