missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dihydro-1,4-benzodioxin-5-ylmethylamine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC01413DE
| Quantity | 5g |
|---|---|
| Packaging | Amber Glass Bottle |
Chemical Identifiers
| 261633-71-0 | |
| 165.192 | |
| 2,3-dihydrobenzo b 1,4 dioxin-5-yl methanamine, c-2,3-dihydro-benzo 1,4 dioxin-5-yl-methylamine, 1,4-benzodioxin-5-methanamine, 2,3-dihydro, 2,3-dihydro-1,4-benzodioxin-5-ylmethylamine, 5-aminomethyl-1,4-benzodioxane, 1-2,3-dihydro-1,4-benzodioxin-5-yl methanamine, c-2,3-dihydrobenzo 1,4 dioxin-5-yl methylamine | |
| 2,3-dihydro-1,4-benzodioxin-5-ylmethanamine |
| C9H11NO2 | |
| OKRXRKUGZVMPKX-UHFFFAOYSA-N | |
| 2795040 | |
| C1COC2=C(C=CC=C2O1)CN |
Specifications
| 2, 3-Dihydro-1, 4-benzodioxin-5-ylmethylamine hydrochloride | |
| 95% | |
| C9H11NO2 | |
| 2,3-dihydrobenzo b 1,4 dioxin-5-yl methanamine, c-2,3-dihydro-benzo 1,4 dioxin-5-yl-methylamine, 1,4-benzodioxin-5-methanamine, 2,3-dihydro, 2,3-dihydro-1,4-benzodioxin-5-ylmethylamine, 5-aminomethyl-1,4-benzodioxane, 1-2,3-dihydro-1,4-benzodioxin-5-yl methanamine, c-2,3-dihydrobenzo 1,4 dioxin-5-yl methylamine | |
| C1COC2=C(C=CC=C2O1)CN | |
| 165.192 | |
| 201.65 |
| 261633-71-0 | |
| Amber Glass Bottle | |
| 5g | |
| OKRXRKUGZVMPKX-UHFFFAOYSA-N | |
| 2,3-dihydro-1,4-benzodioxin-5-ylmethanamine | |
| 2795040 | |
| 95% |