missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Difluorophenol 98.0+%, TCI America™
$65.24 - $281.48
Chemical Identifiers
| CAS | 6418-38-8 |
|---|---|
| Molecular Formula | C6H4F2O |
| Molecular Weight (g/mol) | 130.094 |
| MDL Number | MFCD00010262 |
| InChI Key | RPEPGIOVXBBUMJ-UHFFFAOYSA-N |
| Synonym | phenol, 2,3-difluoro, phenol, difluoro, 2,3-difluoro phenol, difluorophenol, difluoro-phenol, 2.3-difluorophenol, 2,3-difluoro-phenol, pubchem1494, acmc-1b6pt, 2,3-bis fluoranyl phenol |
| PubChem CID | 80879 |
| IUPAC Name | 2,3-difluorophenol |
| SMILES | C1=CC(=C(C(=C1)F)F)O |
Chemical Identifiers
| 6418-38-8 | |
| 130.094 | |
| RPEPGIOVXBBUMJ-UHFFFAOYSA-N | |
| 80879 | |
| C1=CC(=C(C(=C1)F)F)O |
| C6H4F2O | |
| MFCD00010262 | |
| phenol, 2,3-difluoro, phenol, difluoro, 2,3-difluoro phenol, difluorophenol, difluoro-phenol, 2.3-difluorophenol, 2,3-difluoro-phenol, pubchem1494, acmc-1b6pt, 2,3-bis fluoranyl phenol | |
| 2,3-difluorophenol |
Specifications
| 6418-38-8 | |
| White | |
| C6H4F2O | |
| 5 g | |
| phenol, 2,3-difluoro, phenol, difluoro, 2,3-difluoro phenol, difluorophenol, difluoro-phenol, 2.3-difluorophenol, 2,3-difluoro-phenol, pubchem1494, acmc-1b6pt, 2,3-bis fluoranyl phenol | |
| C1=CC(=C(C(=C1)F)F)O | |
| 130.094 | |
| 130.09 | |
| Crystalline Powder |
| 40°C | |
| 54°C | |
| MFCD00010262 | |
| 1325 | |
| RPEPGIOVXBBUMJ-UHFFFAOYSA-N | |
| 2,3-difluorophenol | |
| 80879 | |
| ≥98.0% (GC) | |
| 2,3-Difluorophenol |
Safety and Handling
TSCA : No