missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorophenol 98.0+%, TCI America™

Catalog No. D24535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24535G 5 g
D245325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24535G Supplier TCI America Supplier No. D24535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6418-38-8
Molecular Formula C6H4F2O
Molecular Weight (g/mol) 130.094
MDL Number MFCD00010262
InChI Key RPEPGIOVXBBUMJ-UHFFFAOYSA-N
Synonym phenol, 2,3-difluoro, phenol, difluoro, 2,3-difluoro phenol, difluorophenol, difluoro-phenol, 2.3-difluorophenol, 2,3-difluoro-phenol, pubchem1494, acmc-1b6pt, 2,3-bis fluoranyl phenol
PubChem CID 80879
IUPAC Name 2,3-difluorophenol
SMILES C1=CC(=C(C(=C1)F)F)O

Specifications

Melting Point 40°C
Color White
Boiling Point 54°C
Quantity 5 g
UN Number 1325
Formula Weight 130.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Difluorophenol