missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorophenol, 98+%

CAS: 6418-38-8 | C6H4F2O | 130.094 g/mol

$38.67 - $115.79

Chemical Identifiers

CAS 6418-38-8
Molecular Formula C6H4F2O
Molecular Weight (g/mol) 130.094
MDL Number MFCD00010262
InChI Key RPEPGIOVXBBUMJ-UHFFFAOYSA-N
Synonym phenol, 2,3-difluoro, phenol, difluoro, 2,3-difluoro phenol, difluorophenol, difluoro-phenol, 2.3-difluorophenol, 2,3-difluoro-phenol, pubchem1494, acmc-1b6pt, 2,3-bis fluoranyl phenol
PubChem CID 80879
IUPAC Name 2,3-difluorophenol
SMILES C1=CC(=C(C(=C1)F)F)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1697103
View Documents
Thermo Scientific Chemicals
A1697103
1 g
Each for $38.67
Only null left
 
AAA1697106
View Documents
Thermo Scientific Chemicals
A1697106
5 g
Each for $115.79
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6418-38-8
130.094
RPEPGIOVXBBUMJ-UHFFFAOYSA-N
80879
C1=CC(=C(C(=C1)F)F)O
C6H4F2O
MFCD00010262
phenol, 2,3-difluoro, phenol, difluoro, 2,3-difluoro phenol, difluorophenol, difluoro-phenol, 2.3-difluorophenol, 2,3-difluoro-phenol, pubchem1494, acmc-1b6pt, 2,3-bis fluoranyl phenol
2,3-difluorophenol

Specifications

6418-38-8
53°C to 55°C (25 mmHg)
Phenol-like
MFCD00010262
UN2921
phenol, 2,3-difluoro, phenol, difluoro, 2,3-difluoro phenol, difluorophenol, difluoro-phenol, 2.3-difluorophenol, 2,3-difluoro-phenol, pubchem1494, acmc-1b6pt, 2,3-bis fluoranyl phenol
C1=CC(=C(C(=C1)F)F)O
130.094
130.09
2,3-Difluorophenol
34°C to 38°C
56°C (132°F)
C6H4F2O
1 g
2082265
RPEPGIOVXBBUMJ-UHFFFAOYSA-N
2,3-difluorophenol
80879
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H314-H318-H228
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.

P210-P240-P241-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H228-H302+H312+H332-H314-H335

DOTInformation : Hazard Class: 4.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only