missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorophenol, 98+%

Catalog No. AAA1697106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1697106 5 g
AAA1697103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1697106 Supplier Thermo Scientific Chemicals Supplier No. A1697106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6418-38-8 | C6H4F2O | 130.094 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6418-38-8
Molecular Formula C6H4F2O
Molecular Weight (g/mol) 130.094
MDL Number MFCD00010262
InChI Key RPEPGIOVXBBUMJ-UHFFFAOYSA-N
Synonym phenol, 2,3-difluoro, phenol, difluoro, 2,3-difluoro phenol, difluorophenol, difluoro-phenol, 2.3-difluorophenol, 2,3-difluoro-phenol, pubchem1494, acmc-1b6pt, 2,3-bis fluoranyl phenol
PubChem CID 80879
IUPAC Name 2,3-difluorophenol
SMILES C1=CC(=C(C(=C1)F)F)O

Specifications

Melting Point 34°C to 38°C
Boiling Point 53°C to 55°C (25 mmHg)
Flash Point 56°C (132°F)
Odor Phenol-like
Quantity 5 g
UN Number UN2921
Beilstein 2082265
Formula Weight 130.09
Percent Purity ≥98%
Chemical Name or Material 2,3-Difluorophenol
Hazard Category H228-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318-H228
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Precautionary Statement P210-P240-P241-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only