missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Difluorobenzyl Alcohol 98.0+%, TCI America™
$107.80 - $437.02
Chemical Identifiers
| CAS | 75853-18-8 |
|---|---|
| Molecular Formula | C7H6F2O |
| Molecular Weight (g/mol) | 144.121 |
| MDL Number | MFCD00010312 |
| InChI Key | JSFGDUIJQWWBGY-UHFFFAOYSA-N |
| Synonym | 2,3-difluorobenzyl alcohol, 2,3-difluorophenyl methanol, 2,3-difluorobenzylalcohol, 2,3-difluorophenylalcohol, benzenemethanol, 2,3-difluoro, 2,3-difluorophenyl methan-1-ol, pubchem7358, 2,3-diflorobenzyl alcohol, 2,3difluorophenyl methanol, 2,3 difluorophenyl methanol |
| PubChem CID | 447153 |
| ChEBI | CHEBI:41883 |
| IUPAC Name | (2,3-difluorophenyl)methanol |
| SMILES | C1=CC(=C(C(=C1)F)F)CO |
Chemical Identifiers
| 75853-18-8 | |
| 144.121 | |
| JSFGDUIJQWWBGY-UHFFFAOYSA-N | |
| 447153 | |
| (2,3-difluorophenyl)methanol |
| C7H6F2O | |
| MFCD00010312 | |
| 2,3-difluorobenzyl alcohol, 2,3-difluorophenyl methanol, 2,3-difluorobenzylalcohol, 2,3-difluorophenylalcohol, benzenemethanol, 2,3-difluoro, 2,3-difluorophenyl methan-1-ol, pubchem7358, 2,3-diflorobenzyl alcohol, 2,3difluorophenyl methanol, 2,3 difluorophenyl methanol | |
| CHEBI:41883 | |
| C1=CC(=C(C(=C1)F)F)CO |
Specifications
| 75853-18-8 | |
| C7H6F2O | |
| 5 g | |
| JSFGDUIJQWWBGY-UHFFFAOYSA-N | |
| (2,3-difluorophenyl)methanol | |
| 447153 | |
| 144.12 | |
| Liquid |
| Yellow | |
| MFCD00010312 | |
| 2,3-difluorobenzyl alcohol, 2,3-difluorophenyl methanol, 2,3-difluorobenzylalcohol, 2,3-difluorophenylalcohol, benzenemethanol, 2,3-difluoro, 2,3-difluorophenyl methan-1-ol, pubchem7358, 2,3-diflorobenzyl alcohol, 2,3difluorophenyl methanol, 2,3 difluorophenyl methanol | |
| C1=CC(=C(C(=C1)F)F)CO | |
| 144.121 | |
| CHEBI:41883 | |
| ≥98.0% (GC) | |
| 2,3-Difluorobenzyl Alcohol |
Safety and Handling
TSCA : No