missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorobenzyl Alcohol 98.0+%, TCI America™

Catalog No. D46195G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46195G 5 g
D461925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46195G Supplier TCI America Supplier No. D46195G
Only null left

Chemical Identifiers

CAS 75853-18-8
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.121
MDL Number MFCD00010312
InChI Key JSFGDUIJQWWBGY-UHFFFAOYSA-N
Synonym 2,3-difluorobenzyl alcohol, 2,3-difluorophenyl methanol, 2,3-difluorobenzylalcohol, 2,3-difluorophenylalcohol, benzenemethanol, 2,3-difluoro, 2,3-difluorophenyl methan-1-ol, pubchem7358, 2,3-diflorobenzyl alcohol, 2,3difluorophenyl methanol, 2,3 difluorophenyl methanol
PubChem CID 447153
ChEBI CHEBI:41883
IUPAC Name (2,3-difluorophenyl)methanol
SMILES C1=CC(=C(C(=C1)F)F)CO

Specifications

Color Yellow
Quantity 5 g
Formula Weight 144.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Difluorobenzyl Alcohol