missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Difluorobenzonitrile 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 188°C |
| Quantity | 5 g |
| UN Number | 3276 |
| Formula Weight | 139.10 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,3-Difluorobenzonitrile |
Safety and Handling
| TSCA | No |