missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorobenzonitrile 97.0+%, TCI America™

Catalog No. D35001G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35001G 1 g
D35005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. D35001G missing translation for 'mfr' TCI America missing translation for 'supplierNo' D35001G
Only null left

Chemical Identifiers

CAS 21524-39-0
Molecular Formula C7H3F2N
Molecular Weight (g/mol) 139.105
MDL Number MFCD00009976
InChI Key GKPHNZYMLJPYJJ-UHFFFAOYSA-N
Synonym benzonitrile, 2,3-difluoro, 2,3-difluorobenzenecarbonitrile, ncr bf cf, difluorobenzonitrile, pubchem1564, acmc-209flg, 2,3-difluorobenzonitril, 2,3-difluoro-benzonitrile, 2.3-difluoro benzonitrile, ksc493a8b
PubChem CID 88935
IUPAC Name 2,3-difluorobenzonitrile
SMILES C1=CC(=C(C(=C1)F)F)C#N

Specifications

Color Yellow
Boiling Point 188°C
Quantity 1 g
UN Number 3276
Formula Weight 139.10
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Difluorobenzonitrile