missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Difluoro-5-(trifluoromethyl)pyridine 98.0+%, TCI America™
$82.79 - $236.80
Chemical Identifiers
| CAS | 89402-42-6 |
|---|---|
| Molecular Formula | C6H2F5N |
| Molecular Weight (g/mol) | 183.08 |
| MDL Number | MFCD04972837 |
| InChI Key | XIFCGIKPAAZFFS-UHFFFAOYSA-N |
| PubChem CID | 13421547 |
| IUPAC Name | 2,3-difluoro-5-(trifluoromethyl)pyridine |
| SMILES | FC1=C(F)N=CC(=C1)C(F)(F)F |
Chemical Identifiers
| 89402-42-6 | |
| 183.08 | |
| XIFCGIKPAAZFFS-UHFFFAOYSA-N | |
| 2,3-difluoro-5-(trifluoromethyl)pyridine |
| C6H2F5N | |
| MFCD04972837 | |
| 13421547 | |
| FC1=C(F)N=CC(=C1)C(F)(F)F |
Specifications
| 89402-42-6 | |
| 104°C | |
| MFCD04972837 | |
| 2,3-difluoro-5-trifluoromethyl pyridine, 2,3-difluoro-5-trifluoromethyl-pyridine, pyridine,2,3-difluoro-5-trifluoromethyl, pyridine, 2,3-difluoro-5-trifluoromethyl, acmc-20dwi8, 2,3-di-fluoro-5-trifluoromethyl pyridine, alpha,alpha,alpha,5,6-pentafluoro-3-picoline, 2,3-bis fluoranyl-5-trifluoromethyl pyridine | |
| FC1=C(F)N=CC(=C1)C(F)(F)F | |
| 183.08 | |
| 183.08 | |
| Liquid |
| Colorless | |
| C6H2F5N | |
| 1 g | |
| XIFCGIKPAAZFFS-UHFFFAOYSA-N | |
| 2,3-difluoro-5-(trifluoromethyl)pyridine | |
| 13421547 | |
| ≥98.0% (GC) | |
| 2,3-Difluoro-5-(trifluoromethyl)pyridine |
Safety and Handling
TSCA : No