missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-5-(trifluoromethyl)pyridine 98.0+%, TCI America™

Catalog No. D46231G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46231G 1 g
D46235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46231G Supplier TCI America Supplier No. D46231G
Only null left

Chemical Identifiers

CAS 89402-42-6
Molecular Formula C6H2F5N
Molecular Weight (g/mol) 183.08
MDL Number MFCD04972837
InChI Key XIFCGIKPAAZFFS-UHFFFAOYSA-N
PubChem CID 13421547
IUPAC Name 2,3-difluoro-5-(trifluoromethyl)pyridine
SMILES FC1=C(F)N=CC(=C1)C(F)(F)F

Specifications

Color Colorless
Boiling Point 104°C
Quantity 1 g
Synonym 2,3-difluoro-5-trifluoromethyl pyridine, 2,3-difluoro-5-trifluoromethyl-pyridine, pyridine,2,3-difluoro-5-trifluoromethyl, pyridine, 2,3-difluoro-5-trifluoromethyl, acmc-20dwi8, 2,3-di-fluoro-5-trifluoromethyl pyridine, alpha,alpha,alpha,5,6-pentafluoro-3-picoline, 2,3-bis fluoranyl-5-trifluoromethyl pyridine
Formula Weight 183.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Difluoro-5-(trifluoromethyl)pyridine