missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-5-(trifluoromethyl)pyridine, 98%

CAS: 89402-42-6 | C6H2F5N | 183.08 g/mol

$49.59 - $1435.19

Chemical Identifiers

CAS 89402-42-6
Molecular Formula C6H2F5N
Molecular Weight (g/mol) 183.08
MDL Number MFCD04972837
InChI Key XIFCGIKPAAZFFS-UHFFFAOYSA-N
Synonym 2,3-difluoro-5-trifluoromethyl pyridine, 2,3-difluoro-5-trifluoromethyl-pyridine, pyridine,2,3-difluoro-5-trifluoromethyl, pyridine, 2,3-difluoro-5-trifluoromethyl, acmc-20dwi8, 2,3-di-fluoro-5-trifluoromethyl pyridine, alpha,alpha,alpha,5,6-pentafluoro-3-picoline, 2,3-bis fluoranyl-5-trifluoromethyl pyridine
PubChem CID 13421547
IUPAC Name 2,3-difluoro-5-(trifluoromethyl)pyridine
SMILES FC1=C(F)N=CC(=C1)C(F)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2595803
View Documents
Thermo Scientific Chemicals
H2595803
1 g
Each for $49.59
Only null left
 
AAH2595806
View Documents
Thermo Scientific Chemicals
H2595806
5 g
Each for $166.11
Only null left
 
AAH2595814
View Documents
Thermo Scientific Chemicals
H2595814
25 g
Each for $1,435.19
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

89402-42-6
183.08
XIFCGIKPAAZFFS-UHFFFAOYSA-N
13421547
FC1=C(F)N=CC(=C1)C(F)(F)F
C6H2F5N
MFCD04972837
2,3-difluoro-5-trifluoromethyl pyridine, 2,3-difluoro-5-trifluoromethyl-pyridine, pyridine,2,3-difluoro-5-trifluoromethyl, pyridine, 2,3-difluoro-5-trifluoromethyl, acmc-20dwi8, 2,3-di-fluoro-5-trifluoromethyl pyridine, alpha,alpha,alpha,5,6-pentafluoro-3-picoline, 2,3-bis fluoranyl-5-trifluoromethyl pyridine
2,3-difluoro-5-(trifluoromethyl)pyridine

Specifications

89402-42-6
104°C
C6H2F5N
1 g
XIFCGIKPAAZFFS-UHFFFAOYSA-N
2,3-difluoro-5-(trifluoromethyl)pyridine
13421547
98%
1.47
>100°C (212°F)
MFCD04972837
2,3-difluoro-5-trifluoromethyl pyridine, 2,3-difluoro-5-trifluoromethyl-pyridine, pyridine,2,3-difluoro-5-trifluoromethyl, pyridine, 2,3-difluoro-5-trifluoromethyl, acmc-20dwi8, 2,3-di-fluoro-5-trifluoromethyl pyridine, alpha,alpha,alpha,5,6-pentafluoro-3-picoline, 2,3-bis fluoranyl-5-trifluoromethyl pyridine
FC1=C(F)N=CC(=C1)C(F)(F)F
183.08
183.08
2,3-Difluoro-5-(trifluoromethyl)pyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only