missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-4-[(trans-4-propylcyclohexyl)methoxy]anisole 98.0+%, TCI America™

Catalog No. D49091G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49091G 1 g
D49095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49091G Supplier TCI America Supplier No. D49091G
Only null left

Chemical Identifiers

CAS 1373116-00-7
Molecular Formula C17H24F2O2
Molecular Weight (g/mol) 298.37
MDL Number MFCD28976165
InChI Key VPEMEZFAMQJFFP-UHFFFAOYSA-N
Synonym 2,3-Difluoro-1-methoxy-4-[(trans-4-propylcyclohexyl)methoxy]benzene
PubChem CID 57583104
IUPAC Name 2,3-difluoro-1-methoxy-4-[(4-propylcyclohexyl)methoxy]benzene
SMILES CCCC1CCC(COC2=C(F)C(F)=C(OC)C=C2)CC1

Specifications

Melting Point 53°C
Color White
Quantity 1 g
Formula Weight 298.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Difluoro-4-[(trans-4-propylcyclohexyl)methoxy]anisole